CAS RN 83654-05-1
IUPAC Name:
cyclohexylideneamino N-[6-({[(cyclohexylideneamino)oxy]carbonyl}amino)hexyl]carbamate
Synonyms:
cyclohexylideneamino N-[6-({[(cyclohexylideneamino)oxy]carbonyl}amino)hexyl]carbamate
Molecular Weight (g/mol):
394.52
Molecular Formula:
C20H34N4O4
InChi Key:
RXSVYGIGWRDVQC-UHFFFAOYSA-N
SMILES:
O=C(NCCCCCCNC(=O)ON=C1CCCCC1)ON=C1CCCCC1
1
–
3
of
3
results
1
Promotions Available
Thermo Scientific Chemicals RHC-80267, 98%
Thermo Scientific Chemicals RHC-80267, 98%
CAS: 83654-05-1 Molecular Formula: C20H34N4O4 Synonym: 1,6-Bis(cyclohexyloximinocarbonylamino)hexane; U-57908
2
MilliporeSigma™ Calbiochem™ RHC-80267
Selective inhibitor of DAG lipase activity in canine platelets (IC50 = 4 nM) and in a variety of mammalian cells.
3
R&D Systems™ RHC 80267
CAS: 83654-05-1 Molecular Formula: C20H34N4O4