accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SHNO(S)NH3OO

CAS RN 83730-53-4

IUPAC Name: (2S)-2-azaniumyl-4-[butyl(imino)oxo-λ⁶-sulfanyl]butanoate
Synonyms: (2S)-2-ammonio-4-[butyl(imino)oxo-λ⁶-sulfanyl]butanoate
Molecular Weight (g/mol): 222.3
Molecular Formula: C8H18N2O3S
InChi Key: KJQFBVYMGADDTQ-CVSPRKDYSA-N
SMILES: CCCCS(=N)(=O)CC[C@H]([NH3+])C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)