Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ L-Buthionine sulfoximine
GREENER_CHOICESDP

Catalog No. 6954100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-541-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 69-541-00 Supplier R&D Systems™ Supplier No. 6954/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Irreversible γ-glutamylcysteine synthetase (γ-GCS) inhibitor; induces ferroptosis

L-Buthionine sulfoximine is an irreversible γ-glutamylcysteine synthetase (γ-GCS) inhibitor; depletes glutathione levels by inhibiting synthesis. Induces ferroptosis in vitro.

Specifications

Chemical Name or Material (2S)-2-Amino-4-(S-butylsulfonimidoyl)butanoic acid
CAS 83730-53-4
Quantity 100 mg
Solubility Soluble to 100 mM in water and to 100 mM in DMSO
Product Line Tocris
Molecular Formula C8H18N2O3S
Formula Weight 222.3
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.