accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNH2NNClCl

CAS RN 84057-84-1

IUPAC Name: 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
Synonyms: lamotrigine C9H7cl2N5 lamotrigina faes brand of lamotrigine labileno crisomet lamitor LTG lamiktal lamotrigine preparation
Molecular Weight (g/mol): 256.09
Molecular Formula: C9H7Cl2N5
InChi Key: PYZRQGJRPPTADH-UHFFFAOYSA-N
SMILES: NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)