Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lamotrigine
GREENER_CHOICESDP

Catalog No. 161150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-115-0 50 mg
16-111-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-115-0 Supplier R&D Systems™ Supplier No. 1611/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibits glutamate release. Anticonvulsant

Anticonvulsant. Inhibits glutamate release, possibly through inhibition of Na+, K+ and Ca2+ currents. Also blocks heterologously expressed and native α4β2 nAChRs with a similar affinity to Na+ channels.

Water-soluble Salt also available.

Chemical Identifiers

CAS 84057-84-1
Molecular Formula C9H7Cl2N5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 256.09
Purity >99%
Molecular Weight (g/mol) 256.09
Chemical Name or Material Lamotrigine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.