accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 845861-39-4

IUPAC Name: 4-{5-propoxy-1H-pyrrolo[3,2-b]pyridin-3-yl}-1,2,3,6-tetrahydropyridine hydrochloride
Synonyms: 4-{5-propoxy-1H-pyrrolo[3,2-b]pyridin-3-yl}-1,2,3,6-tetrahydropyridine hydrochloride
Molecular Weight (g/mol): 293.8
Molecular Formula: C15H20ClN3O
InChi Key: PIIOXKQIZCVXMD-UHFFFAOYSA-N
SMILES: Cl.CCCOC1=CC=C2NC=C(C2=N1)C1=CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results