Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CP 94253 hydrochloride
GREENER_CHOICESDP

Catalog No. 131750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-175-0 50 mg
13-171-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-175-0 Supplier R&D Systems™ Supplier No. 1317/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective 5-HT1B agonist

Potent, selective 5-HT1B agonist (Ki values are 89, 2, 860, 49 and 1,600 nM for 5-HT1A, 5-HT1B, 5-HT1C, 5-HT1D and 5-HT2 receptors respectively). Centrally active upon systemic administration in vivo.

Chemical Identifiers

CAS 845861-39-4
Molecular Formula C15H20ClN3O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 293.8
Purity >98%
Molecular Weight (g/mol) 293.795
Chemical Name or Material CP 94253 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.