accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNN

CAS RN 848591-90-2

IUPAC Name: 3-{5-methyl-octahydropyrrolo[3,4-c]pyrrol-2-yl}-6-phenylpyridazine
Synonyms: 3-{5-methyl-hexahydropyrrolo[3,4-c]pyrrol-2-yl}-6-phenylpyridazine
Molecular Weight (g/mol): 280.38
Molecular Formula: C17H20N4
InChi Key: GTMRUYCIJSNXGB-UHFFFAOYNA-N
SMILES: CN1CC2CN(CC2C1)C1=NN=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results