accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NNOSOHHO

CAS RN 849662-80-2

IUPAC Name: 2-(4-hydroxyphenyl)-3-{4-[4-(propan-2-yl)piperazin-1-yl]benzoyl}-1-benzothiophen-6-ol
Synonyms: 2-(4-hydroxyphenyl)-3-[4-(4-isopropylpiperazin-1-yl)benzoyl]-1-benzothiophen-6-ol
Molecular Weight (g/mol): 472.6
Molecular Formula: C28H28N2O3S
InChi Key: LQEOPHGPHCWOAC-UHFFFAOYSA-N
SMILES: CC(C)N1CCN(CC1)C1=CC=C(C=C1)C(=O)C1=C(SC2=CC(O)=CC=C12)C1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)