accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 849773-63-3

IUPAC Name: N-hydroxy-3-methyl-2-[N-(propan-2-yloxy)[1,1'-biphenyl]-4-sulfonamido]butanamide
Synonyms: N-hydroxy-2-{N-isopropoxy[1,1'-biphenyl]-4-sulfonamido}-3-methylbutanamide
Molecular Weight (g/mol): 406.5
Molecular Formula: C20H26N2O5S
InChi Key: DGZZVIWCMGVHGV-UHFFFAOYNA-N
SMILES: CC(C)ON(C(C(C)C)C(=O)NO)S(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results