Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ARP 101
GREENER_CHOICESDP

Catalog No. 26225 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-225 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-225 Supplier R&D Systems™ Supplier No. 2622/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of MMP-2

Selective inhibitor of MMP-2 that displays ∽ 600-fold selectivity over MMP-1 (IC50 values are 0.81 and 486 nM respectively).

Chemical Identifiers

CAS 849773-63-3
Molecular Formula C20H26N2O5S

Specifications

Quantity 5 mg
Product Line Tocris
Formula Weight 406.5
Purity >99%
Molecular Weight (g/mol) 406.497
Chemical Name or Material ARP 101
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.