CAS RN 850140-73-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Afatinib Dimaleate
Potent dual specificity EGFR/HER2 inhibitor; active in vivo