CAS RN 850608-87-6
IUPAC Name:
6-(trifluoromethoxy)-1,3-benzothiazol-2-amine hydrochloride
Synonyms:
riluzole hydrochloride
Molecular Weight (g/mol):
270.65
Molecular Formula:
C8H6ClF3N2OS
InChi Key:
QEAOELIJQRYJJS-UHFFFAOYSA-N
SMILES:
Cl.NC1=NC2=CC=C(OC(F)(F)F)C=C2S1
1
–
1
of
1
results
1
R&D Systems™ Riluzole hydrochloride
CAS: 850608-87-6 Molecular Formula: C8H6ClF3N2OS