Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Riluzole hydrochloride
GREENER_CHOICESDP

Catalog No. 076825 Shop All Novus Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-682-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-682-5 Supplier R&D Systems™ Supplier No. 0768/25

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. GABA uptake inhibitor; also inhibits glutamate release and blocks NaV channels

Inhibits glutamate release from presynaptic terminals and increases Na+-dependent glutamate uptake in rat cortical synaptosomes. Blocks voltage-dependent Na+ channels and inhibits GABA uptake by striatal synaptosomes. Also activates TRPC5 channels. Psychotropic agent with anticonvulsant, hypnotic, anxiolytic, anti-ischemic, anesthetic and neuroprotective properties.

Chemical Identifiers

CAS 850608-87-6
Molecular Formula C8H6ClF3N2OS

Specifications

Quantity 25 mg
Product Line Tocris
Formula Weight 270.66
Purity >98%
Molecular Weight (g/mol) 270.654
Chemical Name or Material Riluzole hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.