accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85650-52-8

IUPAC Name: 5-methyl-2,5,19-triazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene
Synonyms: mirtazapine zispin mirtazapine (product) remeron soltab mirtabene mepirzapin celltech brand of mirtazapine remergon mirtazapina remergil
Molecular Weight (g/mol): 265.36
Molecular Formula: C17H19N3
InChi Key: RONZAEMNMFQXRA-UHFFFAOYNA-N
SMILES: CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CN=C21
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)