accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85803-43-6

SOHNH2(R)

CAS RN 85803-43-6

IUPAC Name: (2R)-2-amino-3-(benzylsulfanyl)propan-1-ol
Synonyms: (2R)-2-amino-3-(benzylsulfanyl)propan-1-ol
Molecular Weight (g/mol): 197.3
Molecular Formula: C10H15NOS
InChi Key: AUOITUNWAPLMBM-SNVBAGLBSA-N
SMILES: N[C@H](CO)CSCC1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)