CAS RN 866021-03-6
CAS RN 866021-03-6
IUPAC Name:
1-(4-chlorophenyl)-4-({1H-pyrrolo[2,3-b]pyridin-3-yl}methyl)piperazine trihydrochloride
Synonyms:
1-(4-chlorophenyl)-4-{1H-pyrrolo[2,3-b]pyridin-3-ylmethyl}piperazine trihydrochloride
Molecular Weight (g/mol):
436.2
Molecular Formula:
C18H22Cl4N4
InChi Key:
KJSOYZLYCFYXFC-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.ClC1=CC=C(C=C1)N1CCN(CC2=CNC3=NC=CC=C23)CC1
1
–
1
of
1
results
1
L-745,870 trihydrochloride, Tocris Bioscience™


CAS: 866021-03-6 Molecular Formula: C18H22Cl4N4 Molecular Weight (g/mol): 436.202