accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 868359-05-1

IUPAC Name: 2-amino-3-{[4-(2-bromo-4,5-difluorophenoxy)phenyl]carbamoyl}propanoic acid
Synonyms: 2-amino-3-{[4-(2-bromo-4,5-difluorophenoxy)phenyl]carbamoyl}propanoic acid
Molecular Weight (g/mol): 415.19
Molecular Formula: C16H13BrF2N2O4
InChi Key: BNYDDAAZMBUFRG-UHFFFAOYNA-N
SMILES: NC(CC(=O)NC1=CC=C(OC2=CC(F)=C(F)C=C2Br)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results