CAS RN 868698-49-1
IUPAC Name:
1-(1H-imidazol-5-yl)propan-2-amine dihydrobromide
Synonyms:
α-methylhistamine dihydrobromide
Molecular Weight (g/mol):
287
Molecular Formula:
C6H13Br2N3
InChi Key:
RWHNAAABSGVRDT-UHFFFAOYNA-N
SMILES:
Br.Br.CC(N)CC1=CN=CN1
1
–
1
of
1
results
1
R&D Systems™ (R)-(-)-α-Methylhistamine dihydrobromide
CAS: 868698-49-1 Molecular Formula: C6H13Br2N3