Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (R)-(-)-α-Methylhistamine dihydrobromide
GREENER_CHOICESDP

Catalog No. 056950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-695-0 50 mg
05-691-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 05-695-0 Supplier R&D Systems™ Supplier No. 0569/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, standard H3 agonist

Very potent, high affinity H3 agonist (KD = 50.3 nM) that displays > 200-fold selectivity over H4 receptors. Inhibits H3-mediated histamine synthesis and release in the CNS and stimulates H4-mediated eosinophil shape change (EC50 = 66 nM).

S-enantiomer also available.

Chemical Identifiers

CAS 868698-49-1
Molecular Formula C6H13Br2N3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 287
Purity >99%
Molecular Weight (g/mol) 286.999
Chemical Name or Material (R)-(-)-α-Methylhistamine dihydrobromide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.