CAS RN 871543-07-6
IUPAC Name:
4-methyl-5-[(2-methyl-1,4-diazepan-1-yl)sulfonyl]isoquinoline dihydrochloride
Synonyms:
4-methyl-5-(2-methyl-1,4-diazepan-1-ylsulfonyl)isoquinoline dihydrochloride
Molecular Weight (g/mol):
392.34
Molecular Formula:
C16H23Cl2N3O2S
InChi Key:
BFOPDSJOLUQULZ-UHFFFAOYNA-N
SMILES:
Cl.Cl.CC1CNCCCN1S(=O)(=O)C1=C2C(C)=CN=CC2=CC=C1
1
–
1
of
1
results
1
R&D Systems™ H 1152 dihydrochloride
CAS: 871543-07-6 Molecular Formula: C16H23Cl2N3O2S