Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ H 1152 dihydrochloride
GREENER_CHOICESDP

Catalog No. 24141 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-141 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-141 Supplier R&D Systems™ Supplier No. 2414/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective Rho-kinase (ROCK) inhibitor

Rho-kinase (ROCK) inhibitor that displays high selectivity over other protein kinases (IC50 values are 0.012, 0.180, 0.360, 0.745, 3.03, 5.68 and 28.3 μM for ROCKII, CAMKII, PKG, Aurora A, PKA, PKC and MLCK respectively). Inhibits sulprostone-induced contractions in guinea pig aorta (IC50 = 190 nM) and displays proerectile effects in rats.

Glycyl Derivative also available.

Chemical Identifiers

CAS 871543-07-6
Molecular Formula C16H23Cl2N3O2S

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 392.34
Purity >98%
Molecular Weight (g/mol) 392.339
Chemical Name or Material H 1152 dihydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.