accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3ONNHNOCH3

CAS RN 875634-01-8

IUPAC Name: 5-[3-(2-methoxyphenyl)-1H-pyrrolo[2,3-b]pyridin-5-yl]-N,N-dimethylpyridine-3-carboxamide
Synonyms: 5-[3-(2-methoxyphenyl)-1H-pyrrolo[2,3-b]pyridin-5-yl]-N,N-dimethylpyridine-3-carboxamide
Molecular Weight (g/mol): 372.43
Molecular Formula: C22H20N4O2
InChi Key: GYQRHHQPEMOLKH-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=CNC2=NC=C(C=C12)C1=CC(=CN=C1)C(=O)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)