accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNHNHNNNCH3HClHClHCl

CAS RN 875756-97-1

IUPAC Name: 2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-1,3-benzodiazol-2-yl]-1H-1,3-benzodiazole trihydrochloride
Synonyms: bisbenzimide trihydrochloride
Molecular Weight (g/mol): 561.94
Molecular Formula: C27H31Cl3N6O
InChi Key: JABNPSKWVNCGMX-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.CCOC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NC2=CC=C(C=C2N1)N1CCN(C)CC1
Color 
  • (3)
Label Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)