accessibility menu, dialog, popup

UserName

Viewing profile as user:
KNH2NH2SOOO

CAS RN 88-45-9

IUPAC Name: potassium 2,5-diaminobenzene-1-sulfonate
Synonyms: potassium 2,5-diaminobenzenesulfonate
Molecular Weight (g/mol): 226.29
Molecular Formula: C6H7KN2O3S
InChi Key: WRFPRPDYXWUOEK-UHFFFAOYSA-M
SMILES: [K+].NC1=CC=C(N)C(=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results