Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Phenylenediamine-2-sulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. P16915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16915G 5 g
P169125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16915G Supplier TCI America Supplier No. P16915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88-45-9
Molecular Formula C6H7KN2O3S
Molecular Weight (g/mol) 226.29
MDL Number MFCD00007904
InChI Key WRFPRPDYXWUOEK-UHFFFAOYSA-M
Synonym benzenesulfonic acid, 2,5-diamino, p-phenylenediaminesulfonic acid, 2,5-diaminobenzenesulphonic acid, 2,5-diaminobenzene sulfonic acid, 2-sulfo-p-phenylenediamine, ccris 2884, 1,4-phenylenediamine-2-sulfonic acid, unii-up8112d78n, 2,5-diamino benzene sulfonic acid, pubchem13625
PubChem CID 66617
IUPAC Name potassium 2,5-diaminobenzene-1-sulfonate
SMILES [K+].NC1=CC=C(N)C(=C1)S([O-])(=O)=O

Specifications

Melting Point 300°C
Quantity 5 g
Formula Weight 188.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Phenylenediamine-2-sulfonic Acid
EINECSNumber (3)-2058
RTECSNumber DB5900100
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.