accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 880-78-4

IUPAC Name: 1,2,3,4,5-pentafluoro-6-nitrobenzene
Synonyms: pentafluoronitrobenzene
Molecular Weight (g/mol): 213.06
Molecular Formula: C6F5NO2
InChi Key: INUOFQAJCYUOJR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)