accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSOONH3CCH3CH3

CAS RN 885434-70-8

IUPAC Name: 2,4-dimethyl-1-(2,4,6-trimethylbenzenesulfonyl)-1H-pyrrole
Synonyms: 2,4-dimethyl-1-(2,4,6-trimethylbenzenesulfonyl)pyrrole
Molecular Weight (g/mol): 277.38
Molecular Formula: C15H19NO2S
InChi Key: AFZWZVLPIMHLSE-UHFFFAOYSA-N
SMILES: CC1=CN(C(C)=C1)S(=O)(=O)C1=C(C)C=C(C)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)