CAS RN 887401-93-6
IUPAC Name:
N-{1-[4-hydroxy-4-(6-methoxypyridin-3-yl)cyclohexyl]pyrrolidin-3-yl}-2-{[3-(trifluoromethyl)phenyl]formamido}acetamide; bis(methanesulfonic acid)
Synonyms:
N-{1-[4-hydroxy-4-(6-methoxypyridin-3-yl)cyclohexyl]pyrrolidin-3-yl}-2-{[3-(trifluoromethyl)phenyl]formamido}acetamide; bis(methanesulfonic acid)
Molecular Weight (g/mol):
712.75
Molecular Formula:
C28H39F3N4O10S2
InChi Key:
PNPNUHKICDECDH-UHFFFAOYNA-N
SMILES:
CS(O)(=O)=O.CS(O)(=O)=O.COC1=NC=C(C=C1)C1(O)CCC(CC1)N1CCC(C1)NC(=O)CNC(=O)C1=CC(=CC=C1)C(F)(F)F
1
–
1
of
1
results
1
R&D Systems™ INCB 3284 dimesylate
CAS: 887401-93-6 Molecular Formula: C28H39F3N4O10S2