CAS RN 894804-07-0
IUPAC Name:
N-(4-amino-5-cyano-6-ethoxypyridin-2-yl)-2-(2,5-dimethoxyphenyl)acetamide
Synonyms:
N-(4-amino-5-cyano-6-ethoxypyridin-2-yl)-2-(2,5-dimethoxyphenyl)acetamide
Molecular Weight (g/mol):
356.38
Molecular Formula:
C18H20N4O4
InChi Key:
KQMPRSZTUSSXND-UHFFFAOYSA-N
SMILES:
CCOC1=C(C#N)C(N)=CC(NC(=O)CC2=C(OC)C=CC(OC)=C2)=N1
1
–
2
of
2
results
1
R&D Systems™ TCS JNK 6o
A Tocris product, part of R&D Systems. TCS JNK 6o synthesized to Ancillary Material Grade
2
R&D Systems™ TCS JNK 6o
CAS: 894804-07-0 Molecular Formula: C18H20N4O4