Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TCS JNK 6o
GREENER_CHOICESDP

Catalog No. 322250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-225-0 50 mg
32-2210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-225-0 Supplier R&D Systems™ Supplier No. 3222/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective JNK inhibitor

ATP-competitive c-Jun N-terminal kinase (JNK) inhibitor (IC50 values are 2, 4 and 52 nM for JNK1, JNK2 and JNK3 respectively). Displays > 1000 fold selectivity over other kinases, including ERK2 and p38. Inhibits c-Jun phosphorylation (EC50 = 920 nM) and prevents collagen-induced platelet aggregation in vitro.

Chemical Identifiers

CAS 894804-07-0
Molecular Formula C18H20N4O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 356.38
Purity >98%
Molecular Weight (g/mol) 356.382
Chemical Name or Material TCS JNK 6o
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.