accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNHONHHOClHNFCl

CAS RN 896720-20-0

IUPAC Name: 4-{2-[(2-chloro-4-fluorophenyl)amino]-5-methylpyrimidin-4-yl}-N-[1-(3-chlorophenyl)-2-hydroxyethyl]-1H-pyrrole-2-carboxamide
Synonyms: 4-{2-[(2-chloro-4-fluorophenyl)amino]-5-methylpyrimidin-4-yl}-N-[1-(3-chlorophenyl)-2-hydroxyethyl]-1H-pyrrole-2-carboxamide
Molecular Weight (g/mol): 500.36
Molecular Formula: C24H20Cl2FN5O2
InChi Key: WUTVMXLIGHTZJC-UHFFFAOYNA-N
SMILES: CC1=CN=C(NC2=C(Cl)C=C(F)C=C2)N=C1C1=CNC(=C1)C(=O)NC(CO)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)