CAS RN 89835-80-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isochlortetracycline hydrochloride, 97%, can be used as secondary standard, Thermo Scientific™
CAS: 89835-80-3 Molecular Formula: C22H23ClN2O8·HCl Synonym: ISOCTC