CAS RN 899431-18-6
IUPAC Name:
3-{[5-(2,3-dichlorophenyl)-1H-1,2,3,4-tetrazol-1-yl]methyl}pyridine hydrochloride
Synonyms:
3-{[5-(2,3-dichlorophenyl)-1,2,3,4-tetrazol-1-yl]methyl}pyridine hydrochloride
Molecular Weight (g/mol):
342.61
Molecular Formula:
C13H10Cl3N5
InChi Key:
MBTJFFMIPPMRGR-UHFFFAOYSA-N
SMILES:
Cl.ClC1=CC=CC(C2=NN=NN2CC2=CN=CC=C2)=C1Cl
1
–
1
of
1
results
1
R&D Systems™ A 438079 hydrochloride
CAS: 899431-18-6 Molecular Formula: C13H10Cl3N5