Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Molisch Reagent
SCI_ED

Catalog No. S25754 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25754 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25754
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 90-15-3 , 64-18-6
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 7005
ChEBI CHEBI:10319

Specifications

Assay Percent Range 0.05
Density ∽0.8g/mL
Formula Weight 46.07
Packaging Solution
Quantity 100 mL
Chemical Name or Material Molisch Reagent
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.