accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 9002-12-4

NOHNOOOPOOHOOHONOHNOOHOHOHNH3

CAS RN 9002-12-4

IUPAC Name: {[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}({[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy})phosphinic acid amine
Synonyms: [5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy([5-(2,4-dioxo-3H-pyrimidin-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy)phosphinic acid amine
Molecular Weight (g/mol): 567.4
Molecular Formula: C18H26N5O14P
InChi Key: VPXDHZMLJOJKOX-UHFFFAOYNA-N
SMILES: N.OCC1OC(C(O)C1OP(O)(=O)OCC1OC(C(O)C1O)N1C=CC(=O)NC1=O)N1C=CC(=O)NC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results