CAS RN 90134-00-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Di 4 ANEPPS
Voltage-sensitive probe; used to detect changes in membrane potential in electrophysiology protocols