Learn More
Description
In model phospholipid membranes Ex/Em maxima are 465/635 nm, however spectral properties are highly dependent on membrane environment.
Specifications
Specifications
| Chemical Name or Material | 6-[2-(N,N-Dibutylamino)naphthyl]ethenyl-4 '-pyridinium propanesulfonate |
| CAS | 90134-00-2 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Fluorescent Cell Indicators and Sensors |
| Molecular Formula | C28H36N2O3S |
| Purity | 0.9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

