CAS RN 903576-44-3
IUPAC Name:
1-{3-[3-(4-chlorophenyl)propoxy]propyl}piperidine hydrochloride
Synonyms:
pitolisant
Molecular Weight (g/mol):
332.31
Molecular Formula:
C17H27Cl2NO
InChi Key:
XLFKECRRMPOAQS-UHFFFAOYSA-N
SMILES:
Cl.ClC1=CC=C(CCCOCCCN2CCCCC2)C=C1
1
–
1
of
1
results
1
R&D Systems™ BF 2649 hydrochloride
CAS: 903576-44-3 Molecular Formula: C17H27Cl2NO