accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90389-96-1

IUPAC Name: 3-[(methylamino)methyl]benzonitrile
Synonyms: 3-[(methylamino)methyl]benzonitrile
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: KBRCDVSFCMVQSN-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results