CAS RN 9076-44-2
Chymostatin MP Biomedicals
CAS: 9076-44-2 Molecular Formula: C31H41N7O6 Molecular Weight (g/mol): 607.712 InChI Key: MRXDGVXSWIXTQL-MWVOZXJSSA-N Synonym: chymostatin PubChem CID: 129894295 IUPAC Name: (2S)-2-[[(1S)-1-(2-amino-1,4,5,6-tetrahydropyrimidin-6-yl)-2-[[4-methyl-1-oxo-1-[[(2S)-1-oxo-3-phenylpropan-2-yl]amino]pentan-2-yl]amino]-2-oxoethyl]carbamoylamino]-3-phenylpropanoic acid SMILES: CC(C)CC(C(=O)NC(CC1=CC=CC=C1)C=O)NC(=O)C(C2CCN=C(N2)N)NC(=O)NC(CC3=CC=CC=C3)C(=O)O
MilliporeSigma™ Calbiochem™ Chymostatin
Contains chymostatin A as a major form.