CAS RN 911295-24-4
IUPAC Name:
4-[1-(4-hydroxyphenyl)-4-methyl-5-{4-[2-(piperidin-1-yl)ethoxy]phenyl}-2,3-dihydro-1H-pyrazol-3-ylidene]cyclohexa-2,5-dien-1-one dihydrochloride
Synonyms:
methylpiperidinopyrazole
Molecular Weight (g/mol):
542.5
Molecular Formula:
C29H33Cl2N3O3
InChi Key:
SMPRQALBFACGGO-UHFFFAOYSA-N
SMILES:
Cl.Cl.CC1=C(N(NC1=C1C=CC(=O)C=C1)C1=CC=C(O)C=C1)C1=CC=C(OCCN2CCCCC2)C=C1
1
–
1
of
1
results
1
R&D Systems™ MPP dihydrochloride
CAS: 911295-24-4 Molecular Formula: C29H33Cl2N3O3