CAS RN 913844-45-8
IUPAC Name:
2-amino-1-{3-methyl-4-[(4-methylisoquinolin-5-yl)sulfonyl]-1,4-diazepan-1-yl}ethan-1-one dihydrochloride
Synonyms:
2-amino-1-[3-methyl-4-(4-methylisoquinolin-5-ylsulfonyl)-1,4-diazepan-1-yl]ethanone dihydrochloride
Molecular Weight (g/mol):
449.39
Molecular Formula:
C18H26Cl2N4O3S
InChi Key:
ILDBNQGLZFSHQZ-UHFFFAOYNA-N
SMILES:
Cl.Cl.CC1CN(CCCN1S(=O)(=O)C1=C2C(C)=CN=CC2=CC=C1)C(=O)CN
1
–
1
of
1
results
1
R&D Systems™ Glycyl-H 1152 dihydrochloride
CAS: 913844-45-8 Molecular Formula: C18H26Cl2N4O3S