Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Glycyl-H 1152 dihydrochloride
GREENER_CHOICESDP

Catalog No. 24851 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-851 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-851 Supplier R&D Systems™ Supplier No. 2485/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective Rho-kinase (ROCK) inhibitor. More selective analog of H 1152 dihydrochloride (Cat. No. 2414)

Glycyl analog of the Rho-kinase inhibitor H 1152 dihydrochloride (Cat. No. 2414) that displays improved ROCKII selectivity. IC50 values are 0.0118, 2.35, 2.57, 3.26, > 10 and >10 μM for ROCKII, Aurora A, CAMKII, PKG, PKA and PKC respectively.

Chemical Identifiers

CAS 913844-45-8
Molecular Formula C18H26Cl2N4O3S

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 449.4
Purity >99%
Molecular Weight (g/mol) 449.391
Chemical Name or Material Glycyl-H 1152 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.