CAS RN 914471-09-3
IUPAC Name:
4-{[(3-chloro-4-fluorophenyl)amino](nitroso)methylidene}-4,5-dihydro-1,2,5-oxadiazol-3-amine
Synonyms:
4-{[(3-chloro-4-fluorophenyl)amino](nitroso)methylidene}-5H-1,2,5-oxadiazol-3-amine
Molecular Weight (g/mol):
271.64
Molecular Formula:
C9H7ClFN5O2
InChi Key:
MUVQOOPKPBEAJZ-UHFFFAOYSA-N
SMILES:
NC1=NONC1=C(NC1=CC(Cl)=C(F)C=C1)N=O
1
–
1
of
1
results
1
R&D Systems™ INCB 024360-analog, >99%
CAS: 914471-09-3 Molecular Formula: C9H7ClFN5O2