accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 916734-43-5

IUPAC Name: 2-{[3-(benzenesulfonyl)-5,5-dimethyl-1,3-thiazolidin-4-yl]formamido}-3-[(benzylcarbamoyl)amino]propanoic acid
Synonyms: 2-{[3-(benzenesulfonyl)-5,5-dimethyl-1,3-thiazolidin-4-yl]formamido}-3-[(benzylcarbamoyl)amino]propanoic acid
Molecular Weight (g/mol): 520.62
Molecular Formula: C23H28N4O6S2
InChi Key: XKLHCUGVLCGKKX-UHFFFAOYNA-N
SMILES: CC1(C)SCN(C1C(=O)NC(CNC(=O)NCC1=CC=CC=C1)C(O)=O)S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)