Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TC-I 15
GSA_VAGREENER_CHOICESDP

Catalog No. 452750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-275-0 50 mg
45-271-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-275-0 Supplier R&D Systems™ Supplier No. 4527/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent α2β1 inhibitor; displays antithrombotic activity in vivo

TC-I 15 is a potent α2β1 integrin inhibitor (IC50 values for the inhibition of human platelet adhesion to type I collagen are 12 and 715 nM for platelets under static conditions and under flow, respectively). TC-I 15 displays selectivity for α2β1 over αvβ3, α5β1, α6β1 and αIIbβ3 at concentrations exceeding 1000 nM. TC-I 15 reduces collagen IV production in mesangial cells. Active in vivo; TC-I 15 prevents ferric chloride-induced clot formation in mice.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 916734-43-5
Molecular Formula C23H28N4O6S2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 520.62
Purity >98%
Molecular Weight (g/mol) 520.619
Chemical Name or Material TC-I 15
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.