accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 921938-85-4

H3CHNNNOCH3

CAS RN 921938-85-4

IUPAC Name: methyl({4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-7-yl}methyl)amine
Synonyms: methyl({4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazin-7-yl}methyl)amine
Molecular Weight (g/mol): 193.25
Molecular Formula: C10H15N3O
InChi Key: SMECUBWISXUYSV-UHFFFAOYSA-N
SMILES: CNCC1=CC2=C(N=C1)N(C)CCO2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results