CAS RN 923596-52-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ CDK8/19i
Potent and selective CDK8 and CDK19 inhibitor; maintains pluripotency of mouse PSCs in culture