accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOOHONH2OOOH

CAS RN 926281-37-0

IUPAC Name: 2-amino-4-{[3-(carboxymethyl)phenoxy](methoxy)phosphoryl}butanoic acid
Synonyms: methyl carboxymethylphenyl aminocarboxypropylphosphonate
Molecular Weight (g/mol): 331.26
Molecular Formula: C13H18NO7P
InChi Key: NTFPDEDRMYYPAC-UHFFFAOYNA-N
SMILES: COP(=O)(CCC(N)C(O)=O)OC1=CC=CC(CC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)